Assay Details
Gene Symbol: Uba5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009669.1 | NP_001009669.1 | 7 - 8 | 808 | 152 | |
GenBank mRNA | BC088757.1 | - | 7 - 8 | 808 | 152 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
