Assay Details
Gene Symbol: Lhpp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009706.1 | NP_001009706.1 | 3 - 4 | 477 | 144 | |
XM_008759911.1 | XP_008758133.1 | 3 - 4 | 504 | 144 | ||
XM_008759912.1 | XP_008758134.1 | 3 - 4 | 1862 | 144 | ||
GenBank mRNA | BC088448.1 | - | 3 - 4 | 477 | 144 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
