Assay Details
Gene Symbol: Slc17a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009713.2 | NP_001009713.1 | 3 - 4 | 576 | 171 | |
GenBank mRNA | AY800277.1 | - | 3 - 4 | 575 | 171 | |
BC097482.1 | - | 3 - 4 | 576 | 7459136 | 171 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
