Assay Details
Gene Symbol: Cdc42ep2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009689.1 | NP_001009689.1 | 3 | 1138 | 63 | |
XM_006230806.2 | XP_006230868.1 | 3 | 1527 | 63 | ||
XM_006230807.2 | XP_006230869.1 | 3 | 1482 | 63 | ||
XM_006230809.1 | XP_006230871.1 | 3 | 1290 | 63 | ||
XM_008760140.1 | XP_008758362.1 | 2 | 1308 | 63 | ||
XM_008760141.1 | XP_008758363.1 | 2 | 1306 | 63 | ||
XM_008760142.1 | XP_008758364.1 | 3 | 2011 | 63 | ||
GenBank mRNA | BC087091.1 | - | 3 | 1138 | 7190911 | 63 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
