Assay Details
Gene Symbol: Cdh20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012748.1 | NP_001012766.1 | 7 - 8 | 1404 | 131 | |
GenBank mRNA | AB121033.1 | - | 7 - 8 | 1404 | 131 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
