Assay Details
Gene Symbol: Trim5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014023.1 | NP_001014045.1 | 2 - 3 | 526 | 84 | |
XM_006229912.2 | XP_006229974.1 | 2 - 3 | 1012 | 84 | ||
GenBank mRNA | BC078926.1 | - | 2 - 3 | 526 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
