Assay Details
Gene Symbol: Trim5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014023.1 | NP_001014045.1 | 7 - 8 | 1003 | 91 | |
XM_006229912.2 | XP_006229974.1 | 7 - 8 | 1489 | 91 | ||
GenBank mRNA | BC078926.1 | - | 7 - 8 | 1003 | 91 | |
FQ225508.1 | - | 2 - 3 | 122 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
