Assay Details
Gene Symbol: Slco6d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014270.1 | NP_001014292.1 | 2119 | 173 | ||
XM_006245580.2 | XP_006245642.1 | 2357 | 173 | |||
XM_006245583.2 | XP_006245645.1 | 2096 | 173 | |||
GenBank mRNA | BC083829.1 | - | 2119 | 7112354 | 173 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P079563 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 799457 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 363619 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 799954 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 538134 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 882032 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 906867 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 385654 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 116979 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 191034 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 963675 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 236205 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 331674 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 813718 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 890364 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 910942 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 687231 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 731964 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 416404 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 787535 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 40438 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 648319 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 605816 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 445373 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 360398 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 19706 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 361702 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 839103 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 394649 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 538925 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 971427 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 643550 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 573797 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 97180 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 672656 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 300118 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 405949 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 659214 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 131714 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 53437 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 955948 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 32205 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 617352 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 384090 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 474199 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 244928 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 120336 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 872245 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 896837 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 36299 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 648027 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 197434 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 873973 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 524460 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 205044 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 46129 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 12716 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 17733 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 11160 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 3621 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 4898 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 53129 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 7038 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 119497 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 90174 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 123624 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P079563 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P079563 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P079563 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 15705 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 48844 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 89634 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 92059 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 67008 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 42052 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 71111 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 82251 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 13218 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10930162 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10930150 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10930148 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10930158 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10930156 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10930159 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10930154 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10930157 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10930161 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10930155 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10930149 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10930151 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10930153 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10930152 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10930163 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10930147 | - | - |
Affymetrix Rat Clariom S Assay | PSR0900020875.rn.1 | TC0900002537.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0900020876.rn.1 | TC0900002537.rn.1 | RTA TC |
Affymetrix Rat Gene 1.0 ST Array | 10930147 | - | - |
