Assay Details
Gene Symbol: Tmem80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045202.1 | 1 - 2 | 759 | 61 | ||
NR_045203.1 | 1 - 2 | 759 | 61 | |||
GenBank mRNA | BC083751.1 | - | 1 - 2 | 377 | 61 | |
CB546081.1 | - | 1 - 2 | 106 | 61 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | DEAF1 transcription factor | Deaf1 | Anti-sense | Chr. 1 - 214283787 - 214317466 on NCBI Build Rnor_6.0 | ||
Downstream | EPS8-like 2 | Eps8l2 | 5.8kb | Sense | Chr. 1 - 214327992 - 214353466 on NCBI Build Rnor_6.0 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC083751.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
