Assay Details
Gene Symbol: Fam71f1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024324.1 | NP_001019495.1 | 4 - 5 | 859 | 64 | |
XM_006236223.2 | XP_006236285.1 | 6 - 7 | 1221 | 64 | ||
XM_006236225.2 | XP_006236287.1 | 6 - 7 | 1221 | 64 | ||
GenBank mRNA | BC079297.1 | - | 4 - 5 | 859 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
