Assay Details
Gene Symbol: Osgepl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024787.1 | NP_001019958.1 | 3 - 4 | 732 | 99 | |
XM_006244893.2 | XP_006244955.1 | 3 - 4 | 781 | 99 | ||
XM_006244894.2 | XP_006244956.1 | 3 - 4 | 745 | 99 | ||
GenBank mRNA | BC078974.1 | - | 3 - 4 | 752 | 99 | |
BC097950.1 | - | 3 - 4 | 732 | 99 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
