Assay Details
Gene Symbol: Osgepl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024787.1 | NP_001019958.1 | 5 - 6 | 1218 | 66 | |
XM_006244893.2 | XP_006244955.1 | 5 - 6 | 1267 | 66 | ||
XM_006244894.2 | XP_006244956.1 | 5 - 6 | 1231 | 66 | ||
GenBank mRNA | BC078974.1 | - | 5 - 6 | 1238 | 66 | |
BC097950.1 | - | 5 - 6 | 1218 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
