Assay Details
Gene Symbol: Osgepl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024787.1 | NP_001019958.1 | 6 - 7 | 1292 | 89 | |
XM_006244893.2 | XP_006244955.1 | 6 - 7 | 1341 | 89 | ||
XM_006244894.2 | XP_006244956.1 | 6 - 7 | 1305 | 89 | ||
GenBank mRNA | BC097950.1 | - | 6 - 7 | 1292 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
