Assay Details
Gene Symbol: Gli4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029926.1 | NP_001025097.1 | 2 - 3 | 638 | 61 | |
XM_006241856.2 | XP_006241918.1 | 2 - 3 | 237 | 61 | ||
XM_006241858.1 | XP_006241920.1 | 2 - 3 | 522 | 61 | ||
GenBank mRNA | BC097383.1 | - | 2 - 3 | 638 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
