Assay Details
Gene Symbol: Ptbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271057.1 | NP_001257986.1 | 11 - 12 | 1308 | 86 | |
NM_022516.5 | NP_071961.2 | 10 - 11 | 1230 | 86 | ||
GenBank mRNA | BC061858.1 | - | 11 - 12 | 1303 | 86 | |
X60789.1 | - | 10 - 11 | 1193 | 86 | ||
X60790.1 | - | 7 - 8 | 679 | 86 | ||
X74565.1 | - | 11 - 12 | 1289 | 86 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
