Assay Details
Gene Symbol: Fez2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053600.2 | NP_446052.2 | 2 - 3 | 380 | 83 | |
XM_006239637.2 | XP_006239699.1 | 2 - 3 | 431 | 83 | ||
XM_006239638.2 | XP_006239700.1 | 2 - 3 | 431 | 83 | ||
XM_006239639.2 | XP_006239701.1 | 2 - 3 | 431 | 83 | ||
GenBank mRNA | BC100060.1 | - | 2 - 3 | 361 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
