Assay Details
Gene Symbol: Tor1aip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145092.2 | NP_659560.2 | 1 - 2 | 669 | 71 | |
GenBank mRNA | BC086987.1 | - | 1 - 2 | 669 | 71 | |
U19614.1 | - | 1 - 2 | 539 | 71 | ||
U20286.1 | - | 1 - 2 | 126 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
