Assay Details
Gene Symbol: Tor1aip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145092.2 | NP_659560.2 | 4 - 5 | 847 | 111 | |
XM_006250027.2 | XP_006250089.1 | 4 - 5 | 392 | 111 | ||
XM_006250028.2 | XP_006250090.1 | 4 - 5 | 978 | 111 | ||
GenBank mRNA | BC086987.1 | - | 4 - 5 | 847 | 111 | |
U20286.1 | - | 4 - 5 | 304 | 111 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
