Assay Details
Gene Symbol: Dhrs4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153315.2 | NP_695227.2 | 6 - 7 | 799 | 95 | |
GenBank mRNA | AB062758.1 | - | 6 - 7 | 615 | 95 | |
BC070961.1 | - | 6 - 7 | 799 | 7099044 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
