Assay Details
Gene Symbol: Eef1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175838.1 | NP_787032.1 | 5 - 6 | 850 | 81 | |
GenBank mRNA | BC063162.1 | - | 5 - 6 | 846 | 81 | |
BC072542.1 | - | 5 - 6 | 845 | 81 | ||
BC091297.1 | - | 5 - 6 | 831 | 81 | ||
BC111707.1 | - | 4 - 5 | 814 | 81 | ||
BC128723.1 | - | 5 - 6 | 825 | 81 | ||
L10339.1 | - | 4 - 5 | 785 | 81 | ||
X61043.1 | - | 5 - 6 | 850 | 81 | ||
X63561.1 | - | 4 - 5 | 823 | 81 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
