Assay Details
Gene Symbol: Eef1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175838.1 | NP_787032.1 | 6 - 7 | 1107 | 145 | |
GenBank mRNA | BC063162.1 | - | 6 - 7 | 1103 | 6918979 | 145 |
BC072542.1 | - | 6 - 7 | 1102 | 7103726 | 145 | |
BC091297.1 | - | 6 - 7 | 1088 | 7309009 | 145 | |
BC111707.1 | - | 5 - 6 | 1071 | 7367097 | 145 | |
BC128723.1 | - | 6 - 7 | 1082 | 7312569 | 145 | |
L10339.1 | - | 5 - 6 | 1042 | 145 | ||
M81088.1 | - | 1 - 2 | 268 | 145 | ||
X61043.1 | - | 6 - 7 | 1107 | 145 | ||
X63561.1 | - | 5 - 6 | 1080 | 145 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
