Assay Details
Gene Symbol: Cpsf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012351.2 | NP_001012351.1 | 8 | 757 | 100 | |
XM_006248869.2 | XP_006248931.1 | 8 | 872 | 100 | ||
XM_006248870.2 | XP_006248932.1 | 8 | 797 | 100 | ||
GenBank mRNA | BC089824.1 | - | 8 | 759 | 7309116 | 100 |
Gene Symbol: LOC100360200 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002727934.4 | XP_002727980.1 | 2 | 688 | 100 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
