Assay Details
Gene Symbol: Prkch | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031085.2 | NP_112347.1 | 12 - 13 | 1953 | 76 | |
XM_008764720.1 | XP_008762942.1 | 12 - 13 | 1835 | 76 | ||
GenBank mRNA | BC061763.1 | - | 13 - 14 | 2284 | 76 | |
BC081782.1 | - | 12 - 13 | 1953 | 76 | ||
X68400.1 | - | 12 - 13 | 1794 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
