Assay Details
Gene Symbol: Gnl3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081958.1 | NP_001075427.1 | 11 - 12 | 1085 | 88 | |
GenBank mRNA | CB783942.1 | - | 1 - 2 | 124 | 88 | |
FQ223839.1 | - | 723 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
