Assay Details
Gene Symbol: Gnl3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081958.1 | NP_001075427.1 | 13 - 14 | 1372 | 62 | |
GenBank mRNA | CB783942.1 | - | 3 - 4 | 411 | 62 | |
CB801341.1 | - | 1 - 2 | 182 | 62 | ||
FQ223839.1 | - | 1010 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
