Assay Details
Gene Symbol: Gnl3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081958.1 | NP_001075427.1 | 4 - 5 | 230 | 148 | |
GenBank mRNA | CB544748.1 | - | 4 - 5 | 241 | 148 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
