Assay Details
Gene Symbol: Gnl3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081958.1 | NP_001075427.1 | 6 - 7 | 435 | 76 | |
GenBank mRNA | CA512184.1 | - | 1 - 2 | 59 | 76 | |
FQ223839.1 | - | 73 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
