Assay Details
Gene Symbol: Gnl3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081958.1 | NP_001075427.1 | 7 - 8 | 572 | 81 | |
GenBank mRNA | CA512184.1 | - | 2 - 3 | 196 | 81 | |
FQ223839.1 | - | 210 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
