Assay Details
Gene Symbol: MT-ATP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | NC_001665.ATP6.0 | - | 44 | 107 |

More Information
Additional Information:
NC_001665 (strand:+; 7919..8599); mitochondrially encoded ATP synthase 6
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
