Assay Details
Gene Symbol: Efs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106033.2 | NP_001099503.1 | 4 - 5 | 1517 | 65 | |
GenBank mRNA | BC161942.1 | - | 4 - 5 | 1517 | 9023909 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
