Assay Details
Gene Symbol: Rpa3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106584.1 | NP_001100054.1 | 3 - 4 | 312 | 140 | |
GenBank mRNA | FQ228706.1 | - | 3 - 4 | 300 | 140 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
