Assay Details
Gene Symbol: Cd34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107202.2 | NP_001100672.1 | 4 - 5 | 599 | 66 | |
XM_008769857.1 | XP_008768079.1 | 4 - 5 | 889 | 66 | ||
GenBank mRNA | EU448292.1 | - | 4 - 5 | 599 | 66 | |
EU448293.1 | - | 4 - 5 | 599 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
