Assay Details
Gene Symbol: Rin3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108052.1 | NP_001101522.1 | 2 - 3 | 378 | 86 | |
XM_006240473.2 | XP_006240535.1 | 2 - 3 | 647 | 86 | ||
XM_006240474.2 | XP_006240536.1 | 3 - 4 | 435 | 86 | ||
XM_008764804.1 | XP_008763026.1 | 2 - 3 | 5025 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
