Assay Details
Gene Symbol: Rin3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108052.1 | NP_001101522.1 | 4 - 5 | 537 | 127 | |
XM_006240473.2 | XP_006240535.1 | 4 - 5 | 806 | 127 | ||
XM_006240474.2 | XP_006240536.1 | 5 - 6 | 594 | 127 | ||
XM_008764804.1 | XP_008763026.1 | 4 - 5 | 5184 | 127 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
