Assay Details
Gene Symbol: Rin3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108052.1 | NP_001101522.1 | 6 - 7 | 2332 | 82 | |
XM_006240473.2 | XP_006240535.1 | 6 - 7 | 2601 | 82 | ||
XM_006240474.2 | XP_006240536.1 | 7 - 8 | 2389 | 82 | ||
XM_008764804.1 | XP_008763026.1 | 6 - 7 | 6979 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P070043 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 136981 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 150723 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 297915 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 723079 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 683349 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 90551 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 944930 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 203319 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 396170 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 787266 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 664636 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 393417 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 379273 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 498695 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 704978 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 159155 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 771800 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 605458 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 447278 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 5490 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 204669 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 746161 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 779995 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 925425 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 559469 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 629705 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 162442 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 754753 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 98065 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 959512 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 651927 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 518466 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 903338 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 396875 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 905907 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 861882 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 726515 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 230352 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 62899 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 750591 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 969634 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 398264 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 838018 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 611668 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 731909 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 205043 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 23814 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 576107 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 886665 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 54844 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 357424 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 251787 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 839564 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 101738 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 681431 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 426107 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 942217 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 84038 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 139965 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 643430 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 34771 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 471648 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 933204 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 394289 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 785113 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 134584 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 232738 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 946884 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 618755 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 913941 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 283756 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 572048 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 844701 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 326941 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 837234 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 795070 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 847336 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 305020 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 10019 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 681853 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 33744 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 344273 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 21477 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 8981 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 51938 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 50726 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 66601 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 162595 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 11593 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 62085 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 11701 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 14163 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 150022 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 87908 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 113917 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 47390 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 59339 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P070043 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P070043 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P070043 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P761764 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P142773 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 19609 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 61724 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 103750 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10886454 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10886463 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10886460 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10886458 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10886462 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10886464 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10886457 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10886461 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10886459 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10886453 | - | - |
Rat Genome 230 2.0 Array | 1375020_PM_at | - | - |
Rat Genome 230 2.0 Array | 1375020_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0600009484.rn.1 | TC0600001372.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0600009482.rn.1 | TC0600001372.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0600009485.rn.1 | TC0600001372.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0600009481.rn.1 | TC0600001372.rn.1 | RTA TC |
Rat Expression Set 230 | 1375020_PM_at | - | - |
Rat Expression Set 230 | 1375020_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10886453 | - | - |
Rat Genome U34 Set | rc_AA997773_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21578093 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1368843 | - | - |
