Assay Details
Gene Symbol: Tbc1d25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106955.2 | NP_001100425.1 | 2 - 3 | 291 | 60 | |
GenBank mRNA | BC161862.1 | - | 2 - 3 | 291 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
