Assay Details
Gene Symbol: Pcgf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129882.1 | NP_001123354.1 | 4 - 5 | 571 | 119 | |
XM_006231311.2 | XP_006231373.1 | 4 - 5 | 664 | 119 | ||
XM_008760377.1 | XP_008758599.1 | 3 - 4 | 289 | 119 | ||
GenBank mRNA | BC167067.1 | - | 4 - 5 | 571 | 119 | |
FQ214233.1 | - | 4 - 5 | 552 | 119 | ||
FQ220230.1 | - | 2 - 3 | 162 | 119 | ||
FQ233837.1 | - | 4 - 5 | 505 | 119 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
