Assay Details
Gene Symbol: Pcgf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129882.1 | NP_001123354.1 | 8 - 9 | 968 | 145 | |
XM_006231311.2 | XP_006231373.1 | 8 - 9 | 1061 | 145 | ||
GenBank mRNA | BC167067.1 | - | 8 - 9 | 968 | 7104558 | 145 |
FQ214233.1 | - | 8 - 9 | 949 | 145 | ||
FQ220230.1 | - | 6 - 7 | 559 | 145 | ||
FQ233837.1 | - | 8 - 9 | 903 | 146 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P108284 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P067182 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 103334 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 883296 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 167470 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 168611 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 628792 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 314265 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 372526 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 451224 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 770826 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 556211 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 895382 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 348356 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 764096 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 438842 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 405991 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 955652 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 334522 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 650526 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 319855 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 954959 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 845666 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 694105 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 562063 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 623440 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 207056 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 256962 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 743444 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 117948 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 400822 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 397722 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 325346 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 207607 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 193271 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 378990 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 582256 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 968099 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 756243 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 524328 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 329854 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 803312 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 810006 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 923791 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 394066 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 446377 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 957505 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 27752 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 922988 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 322967 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 683208 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 178054 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 663923 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 534869 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 425462 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 459873 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 283801 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 559307 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 422566 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 458909 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 337292 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 272725 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 96822 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 353048 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 935859 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 744436 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 832626 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 318220 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 754813 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 824680 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 801990 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 634479 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 307793 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 871079 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 783134 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 454066 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 568798 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 7828 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 967630 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 89669 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 89727 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 65770 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 89432 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 41944 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 135435 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 128026 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 174538 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 28919 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 111598 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 73408 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 79556 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 150952 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 102212 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P108284 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P067182 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P108284 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P067182 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P067182 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P108284 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10714969 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10714966 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10714964 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10714965 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10714972 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10714968 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10714970 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10714967 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10714963 | - | - |
Rat Genome 230 2.0 Array | 1377042_PM_at | - | - |
Rat Genome 230 2.0 Array | 1377042_at | - | - |
Affymetrix HT Rat Focus Array | 1377042_at | - | - |
Affymetrix HT Rat Focus Array | 1377042_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0100028620.rn.1 | TC0100003534.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100028621.rn.1 | TC0100003534.rn.1 | RTA TC |
Rat Expression Set 230 | 1377042_PM_at | - | - |
Rat Expression Set 230 | 1377042_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10714963 | - | - |
