Assay Details
Gene Symbol: Ddah2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001165936.1 | NP_001159408.1 | 2 - 3 | 502 | 86 | |
NM_212532.2 | NP_997697.1 | 2 - 3 | 549 | 86 | ||
XM_008772714.1 | XP_008770936.1 | 3 - 4 | 648 | 86 | ||
XM_008772715.1 | XP_008770937.1 | 2 - 3 | 811 | 86 | ||
GenBank mRNA | BC086443.1 | - | 2 - 3 | 450 | 7303579 | 86 |
BC097930.1 | - | 2 - 3 | 539 | 7388716 | 86 | |
CO385374.1 | - | 2 - 3 | 549 | 86 | ||
EV770973.1 | - | 2 - 3 | 502 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
