Assay Details
Gene Symbol: Ddah2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001165936.1 | NP_001159408.1 | 6 - 7 | 943 | 85 | |
NM_212532.2 | NP_997697.1 | 6 - 7 | 990 | 85 | ||
XM_008772714.1 | XP_008770936.1 | 7 - 8 | 1089 | 85 | ||
XM_008772715.1 | XP_008770937.1 | 6 - 7 | 1203 | 85 | ||
GenBank mRNA | BC086443.1 | - | 6 - 7 | 891 | 85 | |
BC097930.1 | - | 6 - 7 | 980 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
