Assay Details
Gene Symbol: Aif1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017196.3 | NP_058892.1 | 2 - 3 | 184 | 115 | |
XM_006256061.2 | XP_006256123.1 | 2 - 3 | 279 | 115 | ||
GenBank mRNA | AB000818.1 | - | 2 - 3 | 182 | 115 | |
FM053942.1 | - | 2 - 3 | 171 | 115 | ||
FQ231499.1 | - | 2 - 3 | 182 | 115 | ||
U17919.1 | - | 2 - 3 | 156 | 115 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
