Assay Details
Gene Symbol: Aif1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017196.3 | NP_058892.1 | 4 - 5 | 295 | 73 | |
XM_006256061.2 | XP_006256123.1 | 3 - 4 | 669 | 73 | ||
XM_006256062.2 | XP_006256124.1 | 4 - 5 | 646 | 73 | ||
XM_006256063.2 | XP_006256125.1 | 4 - 5 | 609 | 73 | ||
XM_006256065.2 | XP_006256127.1 | 6 - 7 | 445 | 73 | ||
GenBank mRNA | AB000818.1 | - | 4 - 5 | 293 | 73 | |
AF299328.1 | - | 1 - 2 | 35 | 73 | ||
BC081822.1 | - | 2 - 3 | 1142 | 73 | ||
D82069.1 | - | 6 - 7 | 450 | 73 | ||
FM053942.1 | - | 4 - 5 | 282 | 73 | ||
FQ231499.1 | - | 4 - 5 | 293 | 73 | ||
U10894.1 | - | 1 - 2 | 49 | 73 | ||
U17919.1 | - | 4 - 5 | 267 | 73 | ||
U33471.1 | - | 2 - 3 | 1319 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
