Assay Details
Gene Symbol: Cdh9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168630.1 | NP_001162101.1 | 5 - 6 | 996 | 100 | |
XM_006232076.2 | XP_006232138.1 | 7 - 8 | 1220 | 100 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
