Assay Details
Gene Symbol: Dcp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170469.1 | NP_001163940.1 | 1 - 2 | 186 | 73 |
Gene Symbol: LOC100912052 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008766780.1 | XP_008765002.1 | 264 | 73 |
Gene Symbol: LOC100912393 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006244683.2 | XP_006244745.1 | 1 - 2 | 199 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
