Assay Details
Gene Symbol: Tor2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007744.1 | NP_001007745.1 | 2 - 3 | 459 | 72 | |
XM_006233981.2 | XP_006234043.1 | 2 - 3 | 1094 | 72 | ||
GenBank mRNA | BC078943.1 | - | 2 - 3 | 459 | 7109352 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
