Assay Details
Gene Symbol: Tuba3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024336.1 | NP_001019507.1 | 1 - 2 | 44 | 57 | |
GenBank mRNA | BC079242.1 | - | 1 - 2 | 44 | 7132665 | 57 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC079242.1,NM_001024336.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
