Assay Details
Gene Symbol: Pdhx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044242.1 | NP_001037707.1 | 1 - 2 | 37 | 89 | |
GenBank mRNA | AY310145.1 | - | 1 - 2 | 37 | 89 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AY310145.1, NM_001044242.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
