Assay Details
Gene Symbol: Pdhx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044242.1 | NP_001037707.1 | 6 - 7 | 822 | 66 | |
GenBank mRNA | AY310145.1 | - | 6 - 7 | 822 | 66 | |
BC091292.1 | - | 3 - 4 | 459 | 66 | ||
FQ232219.1 | - | 849 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
