Assay Details
Gene Symbol: Pdhx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044242.1 | NP_001037707.1 | 9 - 10 | 1187 | 72 | |
GenBank mRNA | AY310145.1 | - | 9 - 10 | 1187 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P097527 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P134750 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 64830 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 43553 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 178080 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 635675 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 226539 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 213506 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 106887 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 455033 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 92409 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 477678 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 127413 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 272257 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 505338 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 486035 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 911663 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 955977 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 23029 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 560374 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 128067 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 131524 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 77690 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 103084 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P134750 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P134750 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P134750 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P247849 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P15209 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 78030 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 73284 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 56603 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10847788 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10847792 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10847793 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10847787 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10847796 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10847794 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10847799 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10847798 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10847795 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10847797 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10847791 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10847789 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10847790 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10847800 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10847786 | - | - |
Rat Genome 230 2.0 Array | 1382234_PM_at | - | - |
Rat Genome 230 2.0 Array | 1398429_at | - | - |
Rat Genome 230 2.0 Array | 1382234_at | - | - |
Rat Genome 230 2.0 Array | 1398429_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1398429_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1398429_at | - | - |
Affymetrix HT Rat Focus Array | 1382234_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1382234_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0300028617.rn.1 | TC0300003764.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0300028616.rn.1 | TC0300003764.rn.1 | RTA TC |
Rat Expression Set 230 | 1398429_PM_at | - | - |
Rat Expression Set 230 | 1398429_at | - | - |
Rat Expression Set 230 | 1382234_PM_at | - | - |
Rat Expression Set 230 | 1382234_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10847786 | - | - |
Rat Genome U34 Set | rc_AA893184_at | - | - |
Rat Genome U34 Set | rc_AI170418_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1365251 | - | - |
