Assay Details
Gene Symbol: Adrb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012492.2 | NP_036624.2 | 1 | 1355 | 84 | |
GenBank mRNA | AY057895.1 | - | 1603 | 84 | ||
AY057896.1 | - | 1 | 1604 | 84 | ||
BC086538.1 | - | 1 | 1355 | 7114502 | 84 | |
J03024.1 | - | 1 | 1257 | 84 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
